Orders LinkContactUsLinkComponent
Phosphate Cell Test, Spectroquant®, MilliporeSigma
Phosphate Cell Test, Spectroquant®, MilliporeSigma
1 pc, Vanadate-molybdate, Prove, Nova 30/60, Multy, Move, 0.5 - 25.0 mg/L PO₄-P, 1.5 - 76.7 mg/L PO₄,
1.1 - 57.3 mg/L P₂O₅, EPS-Moulding, 25
Catalog # EM1.14546.0001
Supplier #:  1145460001
Phosphate Cell Test, Spectroquant®, MilliporeSigma
Catalog # EM1.14546.0001
CAS Number:  
Phosphate Cell Test, Spectroquant®, MilliporeSigma
1 pc, Vanadate-molybdate, Prove, Nova 30/60, Multy, Move, 0.5 - 25.0 mg/L PO₄-P, 1.5 - 76.7 mg/L PO₄,
1.1 - 57.3 mg/L P₂O₅, EPS-Moulding, 25
CAS Number:  
Price
$7,097.90
Log in for availability
Each (1UN) Case de 8 (1UN)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments
  • Very user friendly, validated methods
  • All the necessary reagents for the analysis are contained in the cells for most the test parameters
  • Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
  • The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times.
Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request).
Documents
Certificates
Search by Lot Number
Specifications
Specifications
  • Chemical method:
    Vanadate-molybdate
  • Pack type:
    EPS-Moulding
  • Size:
    1 pc
  • Range:
    0.5 - 25.0 mg/L PO₄-P, 1.5 - 76.7 mg/L PO₄, <br />1.1 - 57.3 mg/L P₂O₅
  • No. of Tests:
    25
  • For Unit:
    Prove, Nova 30/60, Multy, Move
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 