Orders LinkContactUsLinkComponent
Ammonium Test, Spectroquant®, MilliporeSigma
Ammonium Test, Spectroquant®, MilliporeSigma
Ammonium Test, 1 pc, Indophenol blue, Prove, Nova 60, Multy, Move, 0.010 - 3.00 mg/L, Cardboard Box(es), 250
Catalog # EM1.14752.0002
Número de proveedor #:  EM1.14752.0002
Ammonium Test, Spectroquant®, MilliporeSigma
Catalog # EM1.14752.0002
CAS Number:  
Ammonium Test, Spectroquant®, MilliporeSigma
Ammonium Test, 1 pc, Indophenol blue, Prove, Nova 60, Multy, Move, 0.010 - 3.00 mg/L, Cardboard Box(es), 250
CAS Number:  
Price
$6,459.75
Log in Para disponibilidad
Each (1UN) Case de 8 (1UN)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments
  • Highly stable, ready to use reagent mixtures in a user friendly design
  • Free selection of appropriate cell format according to the measuring range
  • Long shelf life up to 3 years - all at room temperature
  • Automatic selection of the correct method with help of AutoSelector barcode identification
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.
Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request).
Documents
Certificates
Buscar por número de lote
Especificaciones
Especificaciones
  • Chemical method:
    Indophenol blue
  • Pack type:
    Cardboard Box(es)
  • Description:
    Ammonium Test
  • Size:
    1 pc
  • Range:
    0.010 - 3.00 mg/L
  • No. of Tests:
    250
  • For Unit:
    Prove, Nova 60, Multy, Move
Especificaciones
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 