AlphaTec® 88-390T Neoprene and Natural Rubber Latex Gloves, Ansell
Proveedor: ANSELL HEALTHCARE PRODUCTS LLC MX
Certificados
Acerca de este artículo
Type 88-390 gloves provide the right mix of chemical protection, dexterity and sensitivity for mid-level applications.
- Excellent chemical resistance
- Pebble embossed design increases grip on wet or dry surfaces
- Durable
The 13-millimeter blend of neoprene and natural rubber latex delivers optimal durability and chemical resistance.
Use in the aerospace, automotive and transportation, chemical, food processing, food service, petrochemical, pharmaceuticals, and general industries. Uses include canning and food processing; detergents and cleaning liquids; handling acids, alkalies, salts, ketones and alcohols; and janitorial and maintenance work.
Certificaciones: Complies with FDA food handling requirements (21 CFR 177.2600).
Información de pedido: This product contains natural rubber latex, which may cause allergic reactions in some individuals.
Embalaje: Gloves are packaged 12 pairs in a bag; 144 in a carton.
Especificaciones
- Catalog No:
- 89089-172
- 32889-654
- Ambidiestro/Específico para cada mano:
- Específico para cada mano
- Específico para cada mano
- Latex/No contiene Latex:
- Latex
- Latex
- Tipo de puño:
- Pinked
- Pinked