Orders LinkContactUsLinkComponent
NIST Traceable pH Buffers with Temperature Table
NIST Traceable pH Buffers with Temperature Table
Certificates
NIST Traceable pH Buffers with Temperature Table
NIST Traceable pH Buffers with Temperature Table
2 Options Available Below
Product Details & Documents
NIST-traceable buffer for pH electrode calibration and verification.
  • Temperature table on bottle
Measurement results depend on accurate calibrations, and accurate calibrations come from high quality solutions. Each standard solution comes with a certificate of analysis, providing traceability to NIST standards.
Product Family Options
Información del producto
  • Pack type
    Size
    pH
    Availability
    Price
  • Bottle
    500 ml
    4.00
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (500ml)
    Each (500ml) - $1,523.52
    $1,523.52
    Each (500ml)
       $1,523.52
    Supplier Number: 63056121
  • Bottle
    500 ml
    7.00
    Discontinued
    Supplier Number: 63056125
  • Bottle
    500 ml
    10.00
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (500ml)
    Each (500ml) - $1,761.57
    $1,761.57
    Each (500ml)
       $1,761.57
    Supplier Number: 63056129