Orders LinkContactUsLinkComponent
Technical Buffer Solutions, Sachets
Technical Buffer Solutions, Sachets
Technical Buffer Solutions, Sachets
Certificates
Technical Buffer Solutions, Sachets
Technical Buffer Solutions, Sachets
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
NIST-traceable buffer for pH electrode calibration and verification.
  • NIST-traceable
  • Ensure freshness
  • Improve data
Also available in multiple packaging options.
Mettler Toledo buffer sachets ensure a fresh portion of buffer is used for each calibration, improving your pH data.
Product Family Options
Información del producto
  • pH Accuracy
    Pack type
    Size
    pH
    Availability
    Price
  • ±0.02
    Sachet, red
    30×20 ml
    4.01
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 30
    Paquete de 30 - $4,618.17
    $4,618.17
    Paquete de 30
       $4,618.17
    Supplier Number: 51302069
  • ±0.02
    Sachet, green
    30×20 ml
    7.00
    Discontinued
    Supplier Number: 51302047
  • ±0.02
    Sachets, blue
    30×20 ml
    10.01
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 30
    Paquete de 30 - $4,569.09
    $4,569.09
    Paquete de 30
       $4,569.09
    Supplier Number: 51302079
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 