ContactUsLinkComponent
AlphaTec® Solvex® 37-185 Chemical resistant gloves, Ansell
AlphaTec® Solvex® 37-185 Chemical resistant gloves, Ansell
Catálogo # 76169-830
undefined
AlphaTec® Solvex® 37-185 Chemical resistant gloves, Ansell
Catálogo # 76169-830
Número de proveedor:  117299
Especificaciones
  • Color del guante:
    Green
  • Tamaño del guante:
    8
  • Ambidiestro/Específico para cada mano:
    Específico para cada mano
  • Desechable / Reutilizable:
    Reutilizable
  • Aprobados para contacto con alimentos:
    +
  • Latex/No contiene Latex:
    No contiene Latex
  • Con polvo/sin polvo:
    Sin polvo
  • Longitud:
    45.7 cm (18")
  • Grosor:
    22 mil.
  • Tipo de puño:
    Puño tipo guante
  • Referencia:
    76169-830
Especificaciones
Acerca de este artículo
High-comfort, chemical-resistant gloves for a wide range of applications.
  • High-performance nitrile compound provides an outstanding combination of chemical resistance and strength
  • Superior snag, puncture, and abrasion protection over rubber or neoprene gloves
  • Suitable for heavy-duty cleaning applications
  • High levels of flexibility, comfort, and dexterity
  • Gauntlet cuff
Type 37-185 Nitrile Gloves are designed to deliver optimal results in wet or dry work environments where chemical resistance is crucial. Gloves are fully reusable, with an unequaled abrasion resistance (ANSI Level 4). They provide superb comfort for the wearer, are free of protein allergens, and will not promote contact dermatitis. No inner cotton flocking, reducing risk of lint contamination and making it ideal for production areas sensitive to the introduction of external impurities. The sandpatch finish gives the glove a smooth surface that decreases indirect costs thanks to fewer rejects of fragile parts.
Certificaciones: Component materials comply with FDA food handling requirements (21 CFR 177.2600).