GsBP®-FFAP Polar, PEG GC Columns, GS-Tek
Catalog # 10499-506
Supplier: GENERAL SEPARATION TECHNOLOGIES
Specifications
- Formato de columna:Capillary
- Espesor de la película:0.25 µm
- Ø int.:0.25 mm
- Longitud:30 m
- Temperature limit:40 to 260/280°C
- Descripción:10499-506
- Referencia:10499-506
- Nº de proveedor:2125-3002
Specifications
About this item
Typical Application: Acrylates, Alcoholic Beverage Standard: Acids & Esters, Aspirin and Ibuprofen in Methanol, Cresylic Acids, Ethoxyethanol, Fatty Acids (Free), Malt Whiskey, Organic Acids.
- Bondable polyethylene glycol
- Acid modified
- High polar
- Equivalent to USP G25 and G35
Ordering information: These columns are suitable replacements for HP-FFAP, DB-FFAP, Rxt-Stabilwa-DA, CP-FFAP.