GC Ferrules, Graphite Vespel®, Trajan Scientific and Medical
Supplier: Trajan Scientific and Medical
Certificates
About this item
Your chromatography analysis does not end with the selection of the GC column
- Composite of graphite and Vespel®
- Mechanically robust
- Hard material
- Long lifetime
- Forms a strong grip with capillary column
These GC ferrules should be used with MS interfaces.
The combination of components selected for your instrument also make an important contribution to successful separations. SGE brings technology and chromatography expertise to improve your chromatography. SGE manufactures an array of connection types for GC use. Poorly defined or selected connections can lead to an increase in dead volume, leaks from cycled temperatures and mismatched tubing sizes.
Certifications: RoHS compliant per Directive 2011/65/EU
Ordering information: A variety of hole sizes are available for 1/16", 1/8" and 1/4" fittings. Ferrules have a 350°C temperature limit. A variety of profiles are available, including short, blank, 2 and 3-hole ferrules. Ferrules are sold in packs of 10.
Frequently Bought Together