Digital 'Pocket' Refractometer, Model PAL-RI, ATAGO®
Catálogo # 97015-896
Proveedor: ATAGO USA INC.
Especificaciones
- Modelo:PAL-BX/RI
- Resolución:0.0001 nD
- Dimensiones:5.5 X 3.1 X 10.9 cm (23/16 x 11/8 x 45/16")
- Measurement temperature range:10 to 75°C (50 to 167°F)
- Clase de protección internacional:IP65
- Temperatura ambiente:10...40 °C (50...104 °F)
- Tiempo de medición:3 seconds
- Referencia:97015-896
- Gama de medición:1.3306 - 1.5284 nD
- Exactitud:± 0.0003 nD
- Peso:100 g (3.52 oz.)
- Nº de proveedor:3852
Especificaciones
Acerca de este artículo
Model PAL-RI is designed for measuring Refractive Index, the measurement will be displayed continuously like an electric newsboard once the sample has been placed
The device features IP65-rated dustproof and water-resistant protection. A sample volume of 0.3mL is required for measurement results.
Información de pedido: One-year manufacturer’s warranty. Supplied with a storage case and two AAA batteries. A calibration certificate is available. Contact your Avantor representative for information.