Wrist Action Variable Speed Shaker, Burrell Scientific
Variable Speed Wrist Action Shaker, Model 95 AA, Accommodates up to 4 vessels, 115 V, 60 Hz, 30.5 cm (12"), 44.5 cm (17.5"), 44.5 cm (17.5"), 31 kg (68 lbs.)
10161-166
: 075-795-04-19
Wrist Action® Variable Speed Shaker has a variable speed motor which will oscillate from 0 450 Osc/Min.This Shaker has a built in timer to operate from 1 to 60 minutes or can be set to continuous operation.
- With the appropriate clamps and supports it can hold a variety of flask, funnels, and specimen jar
- Hold up to ten 125 mL, eight 250/300 mL, or six 500 mL flask
The Wrist Action® Shaker is designed to maintain a constant swirling action as long as required for almost any laboratory application. The adjustment arm controls the amplitude and agitation of shaking from 0 to 15° arc. Side arms and top platforms increase shaking capacity quickly and easily.
: Complies with UL, CSA.
- Description:Variable Speed Wrist Action Shaker, Model 95 AA, Accommodates up to 4 vessels
- Height:30.5 cm (12")
- Length:44.5 cm (17.5")
- Electrical:115 V, 60 Hz
- Weight:31 kg (68 lbs.)
- Width:44.5 cm (17.5")
- Cat. No.:10161-166
- Tilt angle:0 −15°
- Timer:1 − 60 min. or Continuous
- Model:95-AA
- Speed range:450 rpm
- No. of cycles:60
- Max. load:4.5 kg (10 lb.)