Angled Two Neck Round Bottom Flask, Kimble Chase
250 ml, 83 mm, 19/22, 19/22
Catalog # KT295010-0250
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
Número de proveedor #: 295010-0250
Angled Two Neck Round Bottom Flask, Kimble Chase
Catalog # KT295010-0250
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
Jump to:
Product Details & Documents
Distilling round bottom flask with angled side neck
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Two-neck flask with Standard Taper outer joints. Side neck is angled.
Documents
Especificaciones
- Ext.Ø:83 mm
- Capacity:250 ml
- Side ST Joint Size:19/22
- Center Joint:19/22