ContactUsLinkComponent
UniSampler®, Sampler with Hose, Bürkle
UniSampler®, Sampler with Hose, Bürkle
Catálogo # 10217-062
Proveedor:  BURKLE INC
undefined
UniSampler®, Sampler with Hose, Bürkle
Catálogo # 10217-062
Proveedor:  BURKLE INC
Número de proveedor:  5314-2000
Especificaciones
  • Descripción:
    UniSampler with hose
  • Referencia:
    10217-062
  • Nº de proveedor:
    5314-2000
Especificaciones
Acerca de este artículo
Hand operated vacuum pump sampler, equipped with a 2.5 metre long PVC hose. Ideal for deep, narrow and hard to reach spots.
  • Powerful suction for viscous media
  • Easy to disassemble and clean
Universal bottle adapter made of PVC, for bottle necks dia. 24 mm, 32 mm, 39 mm and 50 mm nominal dimension.
Universal bottle adapter made of PVC, for bottle necks dia. 24 mm, 32 mm, 39 mm and 50 mm nominal dimension.