Orders LinkContactUsLinkComponent
ChemoSampler, Bürkle
ChemoSampler, Bürkle
ChemoSampler PP
Catalog # 10217-152
Supplier:   BURKLE INC
Supplier #:  5336-1000
ChemoSampler, Bürkle
Catalog # 10217-152
Supplier:  BURKLE INC
ChemoSampler, Bürkle
ChemoSampler, Bürkle
ChemoSampler PP
Price
$4,752.44
Log in for availability
Each (1UN)
Product Details & Documents
Specially developed for sample collection of aggressive liquids such as acids, alkalis, cleaning agents, (please check chemical resistance).
  • Resistant to acid and alkalis
  • Two bottle sizes available
The sample is directly extracted into the original sample bottle with screw cap. The bottle closure with tamper-evident safety ring ensures reliable sample transfer. The extension rod makes it possible to extract samples to a depth of 2000 mm, e.g. from tanks, silos or tankers. Basic length 1000 mm, extension rod 1000 mm.
Specifications
  • Description:
    ChemoSampler PP
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Extension for ChemoSampler 100 cm (39.37")
    Extension for ChemoSampler 100 cm (39.37")
    76181-722
    Log in for availability
    Each (100cm) - $2,479.66
Product Information
  • Catalog #
    Availability
    Price
  • Sampling Bottle, PE, 500 mL (16.9 oz.)
    Sampling Bottle, PE, 500 mL (16.9 oz.)
    76182-090
    Log in for availability
    Each (500ml) - $195.53
Product Information
  • Catalog #
    Availability
    Price
  • Sampling Bottle, PE, 1000 mL (33.8 oz.)
    Sampling Bottle, PE, 1000 mL (33.8 oz.)
    76182-092
    Log in for availability
    Each (1,000ml) - $212.59