Orders LinkContactUsLinkComponent
ViscoSampler, Bürkle
ViscoSampler, Bürkle
Supplier:   BURKLE INC
ViscoSampler, Bürkle
Supplier:  BURKLE INC
Certificates
ViscoSampler, Bürkle
ViscoSampler, Bürkle
2 Options Available Below
Product Details & Documents
ViscoSampler with powerful suction for viscous media. The media is drawn up and discharged using the suction flask. Specially developed for viscous substances such as slurries, silt, wet clay and soil samples, soil sediments, oils, emulsions, creams etc.
  • No dirt accumulation
  • Disassembling and cleaning is very easy
  • Colour coded handles available
Made of transparent PP or ultra-pure and chemically inert PTFE/FEP for contamination-free sampling. The sampler can be disassembled and the non porous surfaces completely cleaned.
Product Family Options
Información del producto
  • Capacity
    Insertion Depth
    Availability
    Price
  • 10217-346
    ViscoSampler PTFE/FEP, transparent
    ViscoSampler PTFE/FEP, transparent
    160 ml
    60 mm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $23,740.45
    $23,740.45
    Each (1UN)
       $23,740.45
    Supplier Number: 5331-2060
  • 10217-350
    ViscoSampler PTFE/FEP, transparent
    ViscoSampler PTFE/FEP, transparent
    650 ml
    200 mm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $32,766.13
    $32,766.13
    Each (1UN)
       $32,766.13
    Supplier Number: 5331-2200