QualiSampler Set, Bürkle
QualiSampler set PP
Catalog # 10217-130
Supplier: BURKLE INC
Número de proveedor #: 5330-1000
Jump to:
Product Details & Documents
The handy set, which comes complete with carrying case, combines the LiquiSampler and the ViscoSampler with practical accessories
Models in PP or PTFE/ FEP.
Ordering information: Set consists of LiquiSampler 100 cm, ViscoSampler 100 cm, case, 5 sample bottles and cleaning brush.
Especificaciones
- Description:QualiSampler set PP