Orders LinkContactUsLinkComponent
Liquid Sampler, Bürkle
Liquid Sampler, Bürkle
Supplier:   VWR International
Liquid Sampler, Bürkle
Supplier:  VWR International
Certificates
Liquid Sampler, Bürkle
Liquid Sampler, Bürkle
4 Options Available Below
Product Details & Documents
Samplers for thin to viscous liquids.
  • Liquid Sampler, open with thumb: The valve is opened with the thumb to empty the sample
  • Liquid Sampler, place onto bottle: The valve is opened by attaching the sample container to empty the sample
Easy use with one-handed operation. With discharge funnel for safe filling of the sample bottles. Material V4A (1.4404) ; Container Diameter: 32 mm
Documents
Product Family Options
Opened/Closed with thumb
Información del producto
  • Capacity
    Length
    Availability
    Price
  • 50 ml
    46 cm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $13,140.60
    $13,140.60
    Each (1UN)
       $13,140.60
  • 100 ml
    54 cm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $14,228.20
    $14,228.20
    Each (1UN)
       $14,228.20
Opened/Closed semi-automatically
Información del producto
  • Capacity
    Length
    Availability
    Price
  • 50 ml
    45.5 cm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $14,428.43
    $14,428.43
    Each (1UN)
       $14,428.43
  • 100 ml
    53.5 cm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $16,431.54
    $16,431.54
    Each (1UN)
       $16,431.54