Benchtop Draining Racks, Bürkle
Flexible, PVC, with PP rods, 19.6", 19.6"
Catalog # 10217-322
Supplier: VWR International
Jump to:
Product Details & Documents
The stable rods are individually placed into the holes of the draining rack and are clicked into place. Their positions can be changed at any time. Using a combination of rods of three different lengths (60 mm, 100 mm and extension to 150 mm), the draining rack can be adapted to any apparatus which is to be dried.
- Stable bench top rack, can use both sides
- Rods are simply clicked into place
- Collecting pan can be easily removed for emptying
These draining racks are made of PVC.
Documents
Draining racks
Videos
Especificaciones
- Width:19.6"
- Description:Flexible, PVC, with PP rods
- Height:19.6"