Orders LinkContactUsLinkComponent
Burets, Automatic, Dr Schilling Pattern, Schellbach, Bürkle
Burets, Automatic, Dr Schilling Pattern, Schellbach, Bürkle
Supplier:   BURKLE INC
Burets, Automatic, Dr Schilling Pattern, Schellbach, Bürkle
Supplier:  BURKLE INC
Certificates
Burets, Automatic, Dr Schilling Pattern, Schellbach, Bürkle
Burets, Automatic, Dr Schilling Pattern, Schellbach, Bürkle
2 Options Available Below
Product Details & Documents
Burette with Schellbach stripes type Schilling / Bürklewith original Bürkle dosing armature. A universal aid in the laboratory, electroplating industry, water treatment plants, for field analyses, etc. Precise dosing using the press button, exact fine titration with the micro-screw and precise discharge jet.
  • Burette made of borosilicate glass 3.3
  • Shatterproof, resistant to chemicals
  • Blue ring graduations
  • Left or right hand operation possible
  • Sturdy stand
  • Precise discharge jet, allowing very fine drops
 : Tolerances according to DIN ISO 385
Product Family Options
Información del producto
  • Capacity
    Graduations
    Tolerance
    Availability
    Price
  • 10 ml
    0,05 mL
    ±0.030 ml
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $3,397.76
    $3,397.76
    Each (1UN)
       $3,397.76
    Supplier Number: 9695-1010
  • 50 ml
    0,1 mL
    ±0.045 ml
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $3,675.00
    $3,675.00
    Each (1UN)
       $3,675.00
    Supplier Number: 9695-1050