Orders LinkContactUsLinkComponent
Zone Samplers, Bürkle
Zone Samplers, Bürkle
 :   BURKLE INC
Zone Samplers, Bürkle
Zone Samplers, Bürkle
Zone Samplers, Bürkle
For sampling bulk goods, cross-sectional samples can be taken from different sample depths. Suitable for use with a range of sample substrates from fine powder to coarse grain.
  • Easy to clean, removable tip
  • Colour coding (Multi and Uno samplers)
  • Ultra-pure materials: Stainless steel V4A (1.4404)/AISI 316 L, PTFE or aluminium
  • 3 lengths from 550 to 1500 mm, Jumbo up to 2500 mm
  • Samplers have smallest possible gap between inner and outer tubes
Multi sampler has three or five separate chambers for taking samples at multiple depths in one action. Uno sampler has one chamber for taking a sample at one specified depth. All layer sampler has no chambers, three or five inlets and this allows one sample to be taken at multiple depths. Jumbo sampler has no chambers and three, five or seven inlets; with a 50 mm tube diameter this is ideal for larger grain sizes and quantities.
Product Family Options
All Layer Samplers
  • Description
    Length
    Chamber Volume
    Insertion Depth
    No. of Openings
  • All layer sampler, V4A
    55 cm
    70 ml
    430 mm
    3
    Each (1UN)
    Each (1UN) - $37,173.31
    $37,173.31
    Each (1UN)
       $37,173.31
  • All layer sampler, aluminium
    55 cm
    70 ml
    430 mm
    3
    Each (1UN)
    Each (1UN) - $11,791.24
    $11,791.24
    Each (1UN)
       $11,791.24
  • All layer sampler, V4A
    150 cm
    260 ml
    1355 mm
    5
    Each (1UN)
    Each (1UN) - $50,741.75
    $50,741.75
    Each (1UN)
       $50,741.75
Multi Samplers
  • Description
    Length
    Chamber Volume
    Insertion Depth
    No. of Openings
  • Multi sampler V4A/PTFE
    55 cm
    14 ml
    430 mm
    3
    Each (1UN)
    Each (1UN) - $24,288.62
    $24,288.62
    Each (1UN)
       $24,288.62
     : 5316-0055
  • Multi sampler V4A
    55 cm
    14 ml
    430 mm
    3
    Each (1UN)
    Each (1UN) - $451.15
    $451.15
    Each (1UN)
       $451.15
     : 5316-3055
  • Multi sampler aluminum
    85 cm
    17 ml
    710 mm
    3
    Each (1UN)
    Each (1UN) - $14,623.42
    $14,623.42
    Each (1UN)
       $14,623.42
     : 5317-1085
Uno Samplers
  • Description
    Length
    Chamber Volume
    Insertion Depth
    No. of Openings
  • Uno sampler, VA4/PTFE
    85 cm
    17 ml
    710 mm
    1
    Each (1UN)
    Each (1UN) - $30,340.23
    $30,340.23
    Each (1UN)
       $30,340.23
     : 5316-1085
Jumbo Samplers
  • Description
    Length
    Chamber Volume
    Insertion Depth
    No. of Openings
  • All layer-open iiner tube, aluminium
    95 cm
    880 ml
    850 mm
    3
    Each (1UN)
    Each (1UN) - $28,540.07
    $28,540.07
    Each (1UN)
       $28,540.07
     : 5319-1085
  • Multi-closed chambers, Aluminium/PTFE
    95 cm
    390 ml
    850 mm
    3
    Each (1UN)
    Each (1UN) - $41,269.91
    $41,269.91
    Each (1UN)
       $41,269.91
     : 5319-2085
  • Multi-closed chambers, Aluminium/PTFE
    165 cm
    650 ml
    1500 mm
    5
    Each (1UN)
    Each (1UN) - $48,545.09
    $48,545.09
    Each (1UN)
       $48,545.09
     : 5319-2150
  • Multi-closed chambers, Aluminium/PTFE
    265 cm
    910 ml
    2500 mm
    7
    Each (1UN)
    Each (1UN) - $65,453.67
    $65,453.67
    Each (1UN)
       $65,453.67
     : 5319-2250