Especificaciones
- Tipo:CheMobil
- Color:Blue
- Matriz:PMP
- Referencia:10217-296
- Tube Diameter:6 - 11 mm
- Nº de proveedor:8720-2005
Especificaciones
Acerca de este artículo
These paddle wheel flow indicators give a clear visualisation of the flow rates of liquids or gas in tubing. The flow indicator can operate in either direction of flow and can be mounted at any angle. The paddle wheel impeller starts to rotate at about 150 ml/min. Colour coding - red, black or blue.
CheMobil PMP shows reasonable chemical resistance, which makes it possible to use with a great number of fluids. CheMobil is not however appropriate for strong acids, bases and solvents. CheMobil is steam sterilisable. Maximum operating temperature +60 °C.
CheMobil PMP shows reasonable chemical resistance, which makes it possible to use with a great number of fluids. CheMobil is not however appropriate for strong acids, bases and solvents. CheMobil is steam sterilisable. Maximum operating temperature +60 °C.
The flow indicator can operate in either direction of flow and can be mounted at any angle. The paddle wheel impeller starts to rotate at about 150 ml/min. Colour coding - red, black or blue