Orders LinkContactUsLinkComponent
Accessories for Solvent Pump, Bürkle
Accessories for Solvent Pump, Bürkle
Supplier:   BURKLE INC
Accessories for Solvent Pump, Bürkle
Supplier:  BURKLE INC
Certificates
Accessories for Solvent Pump, Bürkle
Accessories for Solvent Pump, Bürkle
2 Options Available Below
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Product Family Options
Product Information
  • For Use With
    Availability
    Price
  • 10217-236
    Anti-Static Set
    Anti-Static Set
    When decanting flammable liquids without the machines and containers involved in the process being sufficiently grounded, electrostatic build-up can occur, which for many substances can result in fire or an explosion; Burkle’s anti-static set ensures easy and reliable grounding; the anti-static set consists of three color-coded connecting copper cables with clamps in a handy plastic case
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $2,854.48
    $2,854.48
    Each (1UN)
       $2,854.48
    Supplier Number: 5602-1000
  • 76121-370
    Thread Adapter, 38 mm to 2" BSP Inner
    Thread Adapter, 38 mm to 2" BSP Inner
    Used to change a 38 mm thread of a canister to a 2" BSP thread; Suitable for US tin foil canisters with an opening of 38 mm
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $3,007.96
    $3,007.96
    Each (1UN)
       $3,007.96
    Supplier Number: 0600-2018
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.