LaboPlast® Pressure Sprayer, Bürkle
Catalog # 10216-878
Supplier: BURKLE INC
Specifications
- Ø:140 mm (5.51")
- Capacidad:1500 ml (50.72 oz.)
- Descripción:Pressure sprayer standard
- Altura:305 mm (12.01") with cap
- Referencia:10216-878
- Nº de proveedor:0309-0100
Specifications
About this item
Pressure sprayer used for disinfection, cleaning, plant protection, thin layer chromatography, technical and sanitary use. The hand pump is used to produce a pressure of up to 4 bar in the container. Spraying take place by pressing a button.
- Media-contacting parts PP, PE, glass-fibre-reinforced PA, V2A, FKM
- Bottle made of HDPE with nozzle diameter 0.6 mm (0.02")
- Adjustable spray jet
- Temperature max. 30 °C