Orders LinkContactUsLinkComponent
OTAL Hand Pumps, Bürkle
Supplier:   BURKLE INC
OTAL Hand Pumps, Bürkle
Supplier:  BURKLE INC
Certificates
OTAL Hand Pumps, Bürkle
OTAL Hand Pumps, Bürkle
3 Options Available Below
Product Details & Documents
Delivery tubes made of PP are resistant against weak acids, caustic solutions and detergents.
Delivery tubes made of PVDF are especially for acids and caustic solutions; not for strongly oxidizing acids and caustic solutions.
Delivery tubes made of stainless steel 304 are for solvents and liquids of VbF Hazard Class AIII (not easily flammable).
Battery pump for safe filling of car batteries with acid or distilled water.
  • Compact, space saving pumps
  • With universal bungs for container openings of 30-70 mm dia.
  • The OTAL mini pump made of PP (model 5005-1000) has a universal bung for container openings of 20-36 mm (0.79-1.42") dia.
Product Family Options
Información del producto
  • Flow rate
    Immersion
    Availability
    Price
  • 10216-952
    OTAL Hand pump, rigid Polypropylene delivery tube, reusable, 10 mm (0.39") tube Ø
    OTAL Hand pump, rigid Polypropylene delivery tube, reusable, 10 mm (0.39") tube Ø
    4 L/min. (0.14 cfm)
    450 mm (17.7") Depth
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $3,200.35
    $3,200.35
    Each (1UN)
       $3,200.35
    Supplier Number: 5005-1000
  • 10216-954
    OTAL Hand pump, rigid Polypropylene delivery tube, reusable, 12 mm (0.4") tube Ø
    OTAL Hand pump, rigid Polypropylene delivery tube, reusable, 12 mm (0.4") tube Ø
    14 L/min. (0.49 cfm)
    700 mm (27.5") Depth
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $4,187.35
    $4,187.35
    Each (1UN)
       $4,187.35
    Supplier Number: 5005-1200
  • 10216-956
    OTAL Hand pump, rigid Polypropylene delivery tube, reusable, 15 mm (0.6") tube Ø
    OTAL Hand pump, rigid Polypropylene delivery tube, reusable, 15 mm (0.6") tube Ø
    20 L/min. (0.7 cfm)
    700 mm (27.5") Depth
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $32.84
    $32.84
    Each (1UN)
       $32.84
    Supplier Number: 5005-1500