Orders LinkContactUsLinkComponent
LaboPlast® Flachmann Compact Jerricans, Bürkle
LaboPlast® Flachmann Compact Jerricans, Bürkle
10 L, Compact Jerrycan with Spigot Compact, Translucent, 125 x 335 x 335 mm (4.91 x 13.19 x 13.19"), 41 mm (1.61") Opening
Catalog # 76183-156
Supplier:   BURKLE INC
Número de proveedor #:  0435-3010
LaboPlast® Flachmann Compact Jerricans, Bürkle
Catalog # 76183-156
Supplier:  BURKLE INC
LaboPlast® Flachmann Compact Jerricans, Bürkle
LaboPlast® Flachmann Compact Jerricans, Bürkle
10 L, Compact Jerrycan with Spigot Compact, Translucent, 125 x 335 x 335 mm (4.91 x 13.19 x 13.19"), 41 mm (1.61") Opening
Price
$3,009.30
Log in Para disponibilidad
Each (10l)
Product Details & Documents
The compact Jerrycan is ideal as a space-saving canister, with available widths of 6.5 cm (2.56") or 12.5 cm (4.91").
  • Available with or without ³/₄" threaded connector
  • Polypropylene, transparent
  • Includes tamper-proof closure
  • Canister without stopcock is autoclavable
  • Equipped with moulded, indestructible liter graduations
Different liquids can be stored and decanted in a small space.
Compact Jerrycan and Stopcock Set
The compact jerrycan and stopcock set consists of the compact jerrycan with glands, in 5 L (1.32 gal.) or 10 L (1.64 gal.) sizes, and the spigot stopcock.
Compact Jerrycan and Compact Stopcock Set
The Compact Stopcock is the ideal addition to the compact jerrycan. It allows for perfect storage and filling when space is at a premium. The space-saving set consists of the compact jerrycan with glands, in 5 L (1.32 gal.) or 10 L (1.64 gal.) sizes, and the spigot compact.
The compact jerrycan can also be stored lying on its side (in a refrigerator). It is very stable due to the ribbed structure and thick sides.
Especificaciones
  • Ø:
    41 mm (1.61") Opening
  • Capacity:
    10 L
  • Color:
    Translucent
  • Description:
    Compact Jerrycan with Spigot Compact
  • Dimensions:
    125 x 335 x 335 mm (4.91 x 13.19 x 13.19")