ContactUsLinkComponent
PYREX® 9 Depression Spot Plate, Corning
PYREX® 9 Depression Spot Plate, Corning
Catalog # 89090-482
Supplier:  Corning
Product available on GSA Advantage®
undefined
PYREX® 9 Depression Spot Plate, Corning
Catalog # 89090-482
Supplier:  Corning
Supplier Number:  7220-85
Product available on GSA Advantage®
Specifications
  • Descripción:
    9 Depression Glass Spot Plates
  • Referencia:
    89090-482
  • Nº de proveedor:
    7220-85
Specifications
About this item
These pressed PYREX® glass plates have 9 concave depressions which is 7 mm deep and has 22 mm outer diameter.
The plate is made of borosilicate glass which has size 85×100 mm.