Orders LinkContactUsLinkComponent
Corning® Lambda™ Plus Multi-Channel Pipettor, Corning®
Corning® Lambda™ Plus Multi-Channel Pipettor, Corning®
 :   CORNING CA
Corning® Lambda™ Plus Multi-Channel Pipettor, Corning®
Corning® Lambda™ Plus Multi-Channel Pipettor, Corning®
Corning® Lambda™ Plus Multi-Channel Pipettor, Corning®
Available in single-, eight-and twelve-channel configurations, all Lambda™ Plus pipettors feature smooth plunger movement and extremely low pipetting forces.
  • Fully autoclavable and UV resistant
  • Designed for universal fit with all common brands of pipette tips
  • Adjustable tip ejector
  • Volume can be set by turning either the thumb wheel or the plunger button
  • Locking mechanism prevents accidental change
The Lambda Plus pipettors have been designed to maintain their calibration, even after repeated use and autoclaving. A key is included with each pipettor for easy in-house calibration.
Certifications: Factory calibrated to strict ISO 8655 standards.
Ordering information: Three-year manufacturer warranty.
  • Catalog No:
  • 89133-478
  • 89133-480
  • 89133-482
  • 89133-484
  • 89133-486
  • 89133-488
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • I-RECs, ISO 14001 - Warsaw
  • I-RECs, ISO 14001 - Warsaw
  • I-RECs, ISO 14001 - Warsaw
  • I-RECs, ISO 14001 - Warsaw
  • I-RECs, ISO 14001 - Warsaw
  • I-RECs, ISO 14001 - Warsaw
Product Family Options
8-Channel
  • Description
    Volume
    For Use With
  • Single-channel pipettor, autoclavable, universal fit
    0.2 - 2 µl
    10 µl tips
    Case (1UN)
    Case (1UN) - $5,620.00
    $5,620.00
    Case (1UN)
       $5,620.00
     : 4070
      Low Manufacturing Impact
    Specifications
    Cat. No.
    89133-478
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Warsaw
      Low Manufacturing Impact
  • Single-channel pipettor, autoclavable, universal fit
    0.5 - 10 µl
    10 µl tips
    Case (1UN)
    Case (1UN) - $5,620.00
    $5,620.00
    Case (1UN)
       $5,620.00
     : 4071
      Low Manufacturing Impact
    Specifications
    Cat. No.
    89133-480
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Warsaw
      Low Manufacturing Impact
  • Single-channel pipettor, autoclavable, universal fit
    2 - 20 µl
    200 µl tips
    Case (1UN)
    Case (1UN) - $5,620.00
    $5,620.00
    Case (1UN)
       $5,620.00
     : 4072
      Low Manufacturing Impact
    Specifications
    Cat. No.
    89133-482
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Warsaw
      Low Manufacturing Impact
  • Single-channel pipettor, autoclavable, universal fit
    10 - 100 µl
    200 µl tips
    Case (1UN)
    Case (1UN) - $5,620.00
    $5,620.00
    Case (1UN)
       $5,620.00
     : 4073
      Low Manufacturing Impact
    Specifications
    Cat. No.
    89133-484
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Warsaw
      Low Manufacturing Impact
  • Single-channel pipettor, autoclavable, universal fit
    20 - 200 µl
    200 µl tips
    Case (1UN)
    Case (1UN) - $7,340.00
    $7,340.00
    Case (1UN)
       $7,340.00
     : 4074
      Low Manufacturing Impact
    Specifications
    Cat. No.
    89133-486
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Warsaw
      Low Manufacturing Impact
  • Single-channel pipettor, autoclavable, universal fit
    100 - 1000 µl
    1000 µl tips
    Case (1UN)
    Case (1UN) - $5,620.00
    $5,620.00
    Case (1UN)
       $5,620.00
     : 4075
      Low Manufacturing Impact
    Specifications
    Cat. No.
    89133-488
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Warsaw
      Low Manufacturing Impact