Acerca de este artículo
Caron Environmental Test Chambers precisely control temperature and humidity conditions. These chambers deliver dependability and high performance, and are loaded with technical innovations such as touchscreen controls, earth-friendly features, and a customer convenient design. The Environmental Test Chambers are designed in accordance with ICH, Q1A Guidelines, making them ideal for pharmaceutical stability drug testing. Other common applications include shelf life studies, stability and package testing, temperature elevation studies, and more.
- Earth-friendly technologies without compromising performance: 115 V units will plug into a standard laboratory electrical outlet
- Temperature and humidity uniformity are excellent for reliable, repeatable results
- Capacitive humidity sensors
- Door handles that are aesthetically pleasing and designed to enhance ergonomics
Accessories and options are not included. Popular options include: Operation down to 0°C (DEFR304 or DEFR305), humidity operation down to 2% RH (DRYR301), water recycling/delivery system (CRSY102).
Designed in accordance with ICH, Q1A Guidelines, making them ideal for pharmaceutical stability drug testing, shelf life studies, stability and package testing, temperature elevation studies, and more.
Especificaciones
- Catalog No:
- 10204-766
- 10204-764
- 10204-776
- 10204-778
- 10204-772
- Intervalo de temperaturas:
- 5...70 °C (41...158 °F)
- 5...70 °C (41...158 °F)
- 5...70 °C (41...158 °F)
- 5...70 °C (41...158 °F)
- 5...70 °C (41...158 °F)
- Environmentally Preferable:
- Water Efficient
- Water Efficient
- Water Efficient
- Water Efficient
- Water Efficient
- Variación de la temperatura:
- ±0.3 °C
- ±0.3 °C
- ±0.3 °C
- ±0.3 °C
- ±0.3 °C
- Control de temperatura:
- ±0.1 °C
- ±0.1 °C
- ±0.1 °C
- ±0.1 °C
- ±0.1 °C
- Temperature sensor:
- 3-Wire RTD
- 3-Wire RTD
- 3-Wire RTD
- 3-Wire RTD
- 3-Wire RTD
- Humidity range:
- 20 to 98% RH
- 20 to 98% RH
- 20 to 98% RH
- 20 to 98% RH
- 20 to 98% RH
- Control de humedad:
- ±2% RH
- ±2% RH
- ±2% RH
- ±2% RH
- ±2% RH
- Acabado:
- Cold Rolled Steel, Powder Coated
- Cold Rolled Steel, Powder Coated
- Cold Rolled Steel, Powder Coated
- Cold Rolled Steel, Powder Coated
- Cold Rolled Steel, Powder Coated
Especificaciones
Información relacionada
FOCUS Industrial Volume 02 Issue 01 VWR Cell Culture Equipments and Instruments
Documentación del producto
TDS Environmental Chambers.pdf