Specifications
- Tamaño del guante:8
- Ambidiestro/Específico para cada mano:Específico para cada mano
- Desechable / Reutilizable:Reutilizable
- Latex/No contiene Latex:Latex
- Longitud:23.2 cm (9⅛")
- Normativa EN:4342
- Tipo de puño:Puño elástico
- Material del revestimiento:Polyethylene
- Calibre:13
- Acabado:Palm Coated
- Referencia:10063-044
- Nº de proveedor:111675
Specifications
About this item
Offering a dynamic mix of protection, durability, and comfort, these gloves provide ANSI Level 2 cut protection and a lightweight ergonomic fit for a balance of safety and precision when working with small, sharp parts.
- Naturally molding gloves
- Excellent resistance to tears and cuts
- Tough coating provides a sure grip
Diverse HyFlex styles provide the best-in-class comfort, protection and dexterity required for the precise manipulation of small parts, controlled operation of power tools and gear, and direct handling of heavy parts and materials
Use in the aerospace, automotive and transportation, construction, electronics, glass, metal fabrication, paper, plastic extrusion, public utilities, transport equipment/trains and boats, and white goods industries. Uses include final and sub-assembly areas when handling metal components; forming, machining and bending of small sharp parts; glass handling and installations; small parts stamping; and wiring.
Gloves mold naturally to the hand and are soft and cool to the skin for all-day comfort. The polyurethane coating provides ANSI level 5 abrasion protection and excellent resistance to tears, while the tough coating provides a sure grip in both light oil and dry applications. The HyFlex 11-644 glove’s exceptional precision, dexterity, and flexibility when handling small, sharp parts increases worker confidence while on the job. Minimal shrinkage and excellent recovery after laundering extends glove life for reduced total cost of ownership.