Orders LinkContactUsLinkComponent
VWR® Shaking Water Baths
VWR® Shaking Water Baths
Supplier:   VWR International
VWR® Shaking Water Baths
Supplier:  VWR International
Certificates
VWR® Shaking Water Baths
2 Options Available Below
Product Details & Documents
The VWR® series of shaking baths offers two bath models with exceptional temperature performance and usability.
  • Excellent temperature uniformity and stability of ±0.1 °C
  • Easy to insert shaking trays, even when loaded
  • Single point user calibration facility
  • User-defined high temperature alarm and cut-off switch
  • Supplied with highly versatile universal shaking tray
  • Advanced run dry/dry start protection
  • VWR® three-year warranty
Units feature soft start and are extremely quiet during operation. The baths are capable of gently mixing or dynamic shaking depending on the application requirements. They incorporate Set and Forget™ technology - fast heat up and excellent temperature accuracy.
Units feature soft start and are extremely quiet during operation. The baths are capable of gently mixing or dynamic shaking depending on the application requirements. They incorporate Set and Forget™ technology - fast heat up and excellent temperature accuracy.
The magnetically coupled, maintenance free shaking mechanism sits below the shaking tray ensuring a larger working area and enabling the user to easily remove the mechanism for cleaning.
Certifications: 120V models are CSA and CE approved.
Ordering information: All models are supplied with a clear polycarbonate lift-off gabled lid and a Universal shaking tray. To accommodate test tubes or microtubes an accessory test tube tray is available with a choice of 8 racks for most commonly used sizes.
Specifications for Product Family
Especificaciones
  • Catalog No:
  • 10128-126
  • 10128-128
  • Model:
  • 12 L
  • 18 L
  • Temperature range:
  • Ambient +5...99°C (Ambient +9...210°F)
  • Ambient +5...99°C (Ambient +9...210°F)
  • Temperature stability:
  • ±0.1°C (±0.18°F)
  • ±0.1°C (±0.18°F)
  • Temperature uniformity:
  • ±0.1°C (±0.18°F)
  • ±0.1°C (±0.18°F)
  • Heating capacity:
  • 0.8 kW
  • 1.05 kW
  • Capacity:
  • 12 L (3.2 gal.)
  • 18 L (4.8 gal.)
  • Linear speed (strokes/min):
  • 20 - 200 strokes/min.
  • 20 - 200 strokes/min.
  • Linear stroke length:
  • 20 mm
  • 20 mm
  • Electrical:
  • 110/120V, 50/60Hz
  • 110/120V, 50/60Hz
  • Int. W×D×H:
  • 23.5×24 cm (9×9"), tray
  • 23.5×42 cm (9×17"), tray
  • Ext. W×D×H:
  • 33.5×36×27.5 cm (13×14×11")
  • 33.5×56.5×27.5 cm (13×22×11")
  • Weight:
  • 9.2 kg (20 lbs.)
  • 11.2 kg (25 lbs.)
Especificaciones
Product Family Options
Información del producto
  • Availability
    Price
  • 10128-126
    VWR® Shaking Water Bath, 12 L
    VWR® Shaking Water Bath, 12 L
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $125,739.75
    $125,739.75
    Each (1UN)
       $125,739.75
    Especificaciones:
    CSA Certified
    More
    Specifications
    Cat. No.
    10128-126
    Model
    12 L
    Temperature range
    Ambient +5...99°C (Ambient +9...210°F)
    Temperature stability
    ±0.1°C (±0.18°F)
    Temperature uniformity
    ±0.1°C (±0.18°F)
    Heating capacity
    0.8 kW
    Capacity
    12 L (3.2 gal.)
    Linear speed (strokes/min)
    20 - 200 strokes/min.
    Linear stroke length
    20 mm
    Electrical
    110/120V, 50/60Hz
    Int. W×D×H
    23.5×24 cm (9×9"), tray
    Ext. W×D×H
    33.5×36×27.5 cm (13×14×11")
    Weight
    9.2 kg (20 lbs.)
    CSA Certified
  • 10128-128
    VWR® Shaking Water Bath, 18 L
    VWR® Shaking Water Bath, 18 L
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $133,368.47
    $133,368.47
    Each (1UN)
       $133,368.47
    Especificaciones:
    CSA Certified
    More
    Specifications
    Cat. No.
    10128-128
    Model
    18 L
    Temperature range
    Ambient +5...99°C (Ambient +9...210°F)
    Temperature stability
    ±0.1°C (±0.18°F)
    Temperature uniformity
    ±0.1°C (±0.18°F)
    Heating capacity
    1.05 kW
    Capacity
    18 L (4.8 gal.)
    Linear speed (strokes/min)
    20 - 200 strokes/min.
    Linear stroke length
    20 mm
    Electrical
    110/120V, 50/60Hz
    Int. W×D×H
    23.5×42 cm (9×17"), tray
    Ext. W×D×H
    33.5×56.5×27.5 cm (13×22×11")
    Weight
    11.2 kg (25 lbs.)
    CSA Certified
Complementary Products
Información del producto
  • Catalog #
    Description
    Probe
    Range
    Availability
    Price
  • 89369-150
    Platinum high-accuracy freezer themometer
    Stainless-steel probe with handle
    −100.00...+199.99 °C (−148.00...+391.99 °F)
    Log in Para disponibilidad
    Each (1UN) - $6,360.63
Accessories
Información del producto
  • Catalog #
    Availability
    Price
  • Replacement Polycarbonate Lid for 18 L and 26 L Baths
    Replacement Polycarbonate Lid for 18 L and 26 L Baths
    97025-628
    Log in Para disponibilidad
    Each (1UN) - $5,135.43