Limited Volume Inserts, WHEATON®
Insert, 0.25 ml, Flat bottom, Glass, No
Catalog # 89235-528
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
Número de proveedor #: 225350-531
Jump to:
Product Details & Documents
For E-Z Vials (12×32 mm) with wide opening.
- Inserts fit nicely into the vials
- Available with or without writing patch
- Glass and polypropylene limited volume inserts
Purchase vials and snap caps separately or together in convenience packs.
Packaging: Packaged in shrink-wrapped trays of 100 for convenience and cleanliness.
Especificaciones
- Cap included:No
- Capacity:0.25 ml
- Description:Insert
- Material:Glass
- Bottom Style:Flat bottom