ContactUsLinkComponent
Stainless Steel Manifolds for Microtest Plates, Drummond
Stainless Steel Manifolds for Microtest Plates, Drummond
Catalog # 53504-250
undefined
Stainless Steel Manifolds for Microtest Plates, Drummond
Catalog # 53504-250
Supplier Number:  3-000-097
Specifications
  • Descripción:
    Straight, 4 Place for 96 Well Plate, 20 mm Centre-to-Centre, Pin Length 17.5 mm
  • Referencia:
    53504-250
  • Nº de proveedor:
    3-000-097
Specifications
About this item
Stainless steel/PTFE manifolds allow multiple test positions to be fed, rinsed, or aspirated quickly and easily.
  • Autoclavable
  • Use for both sample delivery and aspiration operations
  • Supplied complete with cleaning wire for easy cleaning
  • Positive Luer lock connection
  • Pin has O.D. 0.031" and I.D. 0.018"
The autoclavable manifolds work for both sample delivery and aspiration operations. They include a cleaning wire for easy maintenance.