3B Scientific® Ear Chart
Ear Chart, Laminated, 50 cm, 67 cm
470120-056
: 1001500
This anatomical chart details the anatomy of the inner, outer, and middle ear.
- UV Resistant Paper Resists Fading
- Available With Double Sided Lamination To Prevent Curling
The poster presents information about the human ear in colorful detail.
- Width:67 cm
- Description:Ear Chart
- Length:50 cm
- Finish:Laminated