Eisco® Paramecium Models
Paramecium, 1, 25 cm, 39 cm, 9 cm
470136-000
: ZM0016A
Model clearly shows the oral groove and their continuation into the gullet and the other side shows the details of the internal structure. The basic model shows details in a lower relief, while the standard model shows a finer detail level.
- Description:Paramecium
- Height:9 cm
- Length:39 cm
- Width:25 cm
- No. of Parts:1