3B Scientific® Skin Chart
Skin Chart, Laminated, 50 cm, 67 cm
Catalog # 470120-076
Supplier: AMERICAN 3B SCIENTIFIC SE
Número de proveedor #: 1001512
Jump to:
Product Details & Documents
The anatomical detail of the skin is depicted in full color on this anatomical poster.
- UV Resistant Paper Resists Fading
- Available With Double Sided Lamination To Prevent Curling
The anatomy of the human skin, including all skin layers, as well as some common skin pathologies are displayed.
Especificaciones
- Width:67 cm
- Description:Skin Chart
- Length:50 cm
- Finish:Laminated