Order Entry
Mexico
ContactUsLinkComponent
Eisco® Enlarged Upper Triple Root Molar, 2 Parts
Eisco® Enlarged Upper Triple Root Molar, 2 Parts
Catalog # 470136-042
undefined
Eisco® Enlarged Upper Triple Root Molar, 2 Parts
Catalog # 470136-042
Supplier Number:  AM0046

Specifications

  • Descripción:
    2 part triple root molar
  • Altura:
    21 cm
  • Longitud:
    13 cm
  • Referencia:
    470136-042
  • Anchura:
    13 cm
  • Nº de proveedor:
    AM0046
  • No. of Parts:
    2

Specifications

About this item

An enlarged molar separated into two longitudinal sections through the crown, demonstrating the internal and external anatomy of human teeth.

  • Designed by medical professionals and educators
  • Custom molded using the highest quality nontoxic polymers and PVC
  • Includes a key card identifying nine numbered structures
  • Hand-painted by skilled artists

This hand painted molar model is enlarged 15× to show the finest details. It is separated into two longitudinal sections through the crown, two roots and pulp cavity. It comes rod-mounted on a base, with a key card identifying nine structures numbered on the model.