Order Entry
Mexico
Orders LinkContactUsLinkComponent
 

Eisco® Human Ear

Human Ear, 3, 18 cm, 25 cm, 16 cm
  470135-902
 :  AM0038
Eisco® Human Ear
  470135-902
Eisco® Human Ear
Eisco® Human Ear

Human Ear, 3, 18 cm, 25 cm, 16 cm

 
$1,285.61
   
Each (1UN)
 

This Human Ear model demonstrates external, middle, and internal portion of the ear.

  • Five Times Larger than Life Size
  • Hand-Painted
  • Removable Ossicles and Labyrinth
  • Reference Key Included

It dissects into three parts, and the ossicles and labyrinth are removable.

The model includes a reference key card.

 
  • Description:
    Human Ear
  • Height:
    16 cm
  • Length:
    25 cm
  • Width:
    18 cm
  • No. of Parts:
    3