3B Scientific® Spinal Column Sections
Thoracic Spinal Column, 1
Catalog # 470118-946
Supplier: AMERICAN 3B SCIENTIFIC SE
Número de proveedor #: 1000145
Jump to:
Product Details & Documents
These real life replicas of the anatomy of the spinal column are high quality models.
- Includes 3B smart anatomy
Each model shows the regional vertebrae, intervertebral discs, nerves and spinal cord. Each is supplied on a base with a flexible attachment.
Accessories information: For information about 3B smart anatomy, please see the document attached on this page.
Documents
TDS 3B Smart Anatomy
Datasheet
Specifications
- Description:Thoracic Spinal Column
- No. of Parts:1