3B Scientific® Flexible Spine Model With Ribs And Femur Heads
Catalog # 470118-920
Supplier: AMERICAN 3B SCIENTIFIC SE
Specifications
- Descripción:Flexible Spine With Ribs And Femur Heads
- Referencia:470118-920
- Nº de proveedor:A56/2
- No. of Parts:3
Specifications
About this item
Flexible spine with ribs show the interaction of the ribs, spine and associated structures.
- Three years manufacturers warranty
- Includes 3B smart anatomy
This spine and rib model contains the following features: Full pelvis and occipital plate, fully flexible mounting, L3-L4 disc prolapsed, spinal nerve exits, cervical vertebral artery, male pelvis, movable mounted femur heads.
This spine and rib is an anatomically correct model of this section of human anatomy. A stand is not included with this spine and rib model.
Accessories information: A stand is not included with this model and can be purchased separately. For information about 3B smart anatomy, please see the document attached on this page.