ContactUsLinkComponent
Freeze Dry Flasks, Chemglass
Freeze Dry Flasks, Chemglass
Catálogo # 80070-456
Proveedor:  CHEMGLASS INC.
Small Business Enterprise
undefined
Freeze Dry Flasks, Chemglass
Catálogo # 80070-456
Proveedor:  CHEMGLASS INC.
Número de proveedor:  CG-1611-24
Small Business Enterprise
Especificaciones
  • Capacidad:
    600 ml
  • Descripción:
    Freeze Dry Flasks
  • Referencia:
    80070-456
  • Para ser utilizado con:
    Virtis 149088
  • Nº de proveedor:
    CG-1611-24
Especificaciones
Acerca de este artículo
These flat bottom borosilicate flask can be used with Labconco and Virtis freeze dryers.
Wide mouth design provides easy access sample.
Información de pedido: Flasks only, not suppled with rubber top, filter or O-ring.
Precaución: Labconco and Virtis are not related to or affiliated with Chemglass, and these manufacturers have not approved or validated these products for use on their equipment.