Magnetic Stirring Bars, Octagonal,PTFE, Chemglass
Octagon cross-section features a molded-in pivot ring
The PTFE coated Alnico magnet permits magnetic agitation without contamination.
Frequently Bought Together
Octagon cross-section features a molded-in pivot ring
The PTFE coated Alnico magnet permits magnetic agitation without contamination.