ContactUsLinkComponent
Compact Digital Overhead Stirrers, Caframo
Compact Digital Overhead Stirrers, Caframo
Catalog # 77776-540
Supplier:  CAFRAMO LIMITED
Irregular Voltage
undefined
Compact Digital Overhead Stirrers, Caframo
Catalog # 77776-540
Supplier:  CAFRAMO LIMITED
Supplier Number:  BDC2002
Irregular Voltage
Specifications
  • Descripción:
    Compact Digital Stirrer
  • Referencia:
    77776-540
  • Intervalo de velocidad:
    40 - 2002 rpm
  • Horsepower:
    ¹⁄₁₈ hp
  • Volumen máximo:
    25 L
  • Nº de proveedor:
    BDC2002
  • Par de torsión máximo:
    70 N-cm
  • Viscosidad máxima:
    15000 cP
Specifications
About this item
This Compact Digital Overhead Stirrer maximizes productivity, fits into any area of the laboratory including a fume hood or enclosed workstation, and provides accurate and repeatable results every time.
Suitable for a wide range of mixing applications, this Caframo benchtop overhead stirrers is used in formulations, reaction systems, pharmaceuticals, QA testing, and education classrooms.
With the push of a button, the unique Stirlight™ lights up so that solutions can be seen clearly. The Compact Digital stirrer is powered by a brushless DC (BDC) motor. The belt driven transmission is smooth, quiet and maintenance free. The four button touch control (power on/off, speed increase, speed decrease, light on/off) is easy to use.
Ordering information: Clamp, stand, and impellers are available. Chuck (adjustable up to 10 mm or 3/8 in), chuck key, mounting support rod, external power supply, grounded power cord and manual in four languages (English, French, Spanish and German) are included. Three year warranty.
Stands
Product Information