Semicircular Refraction Cell - Acrylic
Catalog # 470136-570
Supplier: UNITED SCIENTIFIC SUPPLIES
Specifications
- Descripción:Semicircular Refraction Cell
- Referencia:470136-570
- Nº de proveedor:RCSC01
Specifications
About this item
Investigate the optical properties of liquids.
- Demonstrate internal reflection and refraction
- Semicircular design allows for each light ray tracing
These clear acrylic refraction cells are excellent for showing refraction and internal reflection in water, and to measure its refractive index. The cells have uniform wall thickness to eliminate distortions. 12 cm diameter x 2.5 cm high.