VWR® Heavy-Duty Wide Mouth Volumetric Flasks with PTFE Stoppers, Class A, Serialized
Catalog # 10124-178
Supplier: VWR International
Specifications
- Capacidad:5 ml
- Referencia:10124-178
- Stopper No.:13
- Tipo de tapón:PTFE
- Certificación:Individual
Specifications
About this item
Heavy-duty wide-mouth volumetric flasks are made to the highest standards to qualify for Class A certification.
- Borosilicate glass fabrication
- Flasks are individually serialized
Made from borosilicate 3.3 expansion glass. Flasks are serialized and supplied with PTFE stopper.
Ordering information: Flasks come with individual certificate of compliance.