Corning's enhanced T-75 Flask features a U-shaped design, which improves usability while maintaining the same environment for cell growth as previous designs
- Non pyrogenic
- Sterile
The U-shaped T-75 flask includes design advances, such as rounded shoulders, which allows for an easier grip and for better access when removing or tightening the cap. The new ergonomic shape also provides the option to use a larger pipet – up to 50 mL – for maximum flexibility, reduces the number of corners and improve cell scraping. Although the enhanced Corning T-75 flask incorporates a number of helpful design improvements, it continues to have the same excellent environment for cell growth and high-quality manufacturing you have come to expect and rely upon from Corning cell culture products.
- Catalog No:
- 10144-862
- 10144-834
- 10144-832
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Certificación ambiental:
- RECs, ISO 14001 - Oneonta
- RECs, ISO 14001 - Oneonta
- RECs, ISO 14001 - Oneonta