n-Hexano
Catalog # EM1.07023.2511
Supplier: MERCK, S.A. DE C.V. SV
CAS Number:
Specifications
- Tipo de env.:Plastic bottle
- Tamaño:2.5 L
- Referencia:EM1.07023.2511
- Nº de proveedor:1.07023.2511
Specifications
About this item
:
:86.18 g/mol
Merck Index:13,04712
Boiling Pt:69 °C (1013 hPa)
Boiling Pt:69 °C (1013 hPa)
Melting Pt:-94,3 °C
Density:0,659 g/cm³ (20 °C)
Flash Pt:–22 °C
Flash Pt:-22 °C
Storage Temperature:Ambiente
EINECS:203-777-6
UN:1208
ADR:3,II
:M-Clarity™ quality level = MQ200
:GHS02
:GHS08
:GHS07
:GHS09
CAS Number:110-54-3
:92112-69-1
Formula:H₃C(CH₂)₄CH₃
MDL Number:MFCD00009520