ContactUsLinkComponent
Whatman™ Carbon Cap™ Carbon Filtration Capsules, Whatman products (Cytiva)
Whatman™ Carbon Cap™ Carbon Filtration Capsules, Whatman products (Cytiva)
  28137-904
 :  Cytiva
Product available on GSA Advantage®
undefined
Whatman™ Carbon Cap™ Carbon Filtration Capsules, Whatman products (Cytiva)
  28137-904
 :  Cytiva
 :  6704-7500
Product available on GSA Advantage®
  • Descripción:
    Carbon Cap 75, heat-fused capsule filter with activated carbon
  • Matriz:
    PP
  • Referencia:
    28137-904
  • Nº de proveedor:
    6704-7500
Carbon cap capsule filters are filled with activated carbon and include a pleated glass fiber filter. They are suitable for adsorption of organic vapors from air, and for removing color, organics, and chlorine from water.
  • Activated carbon acts as an adsorption media for color, organics, and chlorine
  • Pleated glass microfiber filter structure increases surface area, resulting in high efficiency
  • Retains 99.97% of particles greater than 0.3 µm
Large volume chlorine and organic filtration are facilitated by activated carbon, which adsorbs these molecules.
Carbon cap is a filter filled with high-purity, high-efficiency, acid-washed, activated carbon and a High Efficiency Particulate Air (HEPA) filter. It is made specifically for adsorption of organic vapors, chlorine, and decolorizing liquids.