ContactUsLinkComponent
Whatman™ HEPA-CAP Vent Filters
Whatman™ HEPA-CAP Vent Filters
Supplier:  Cytiva
Certificates
undefined
Whatman™ HEPA-CAP Vent Filters
Supplier:  Cytiva
Certificates
Specifications
  • Catalog No:
  • 14230-540
  • 11008-598
  • 28137-884
  • 28137-886
  • Tipo de carcasa:
  • Polypropylene (PP)
  • Polypropylene (PP)
  • Polypropylene (PP)
  • Polypropylene (PP)
  • Presión máx.:
  • 4.1 bar (60 psi)
  • 4.1 bar (60 psi)
  • 4.1 bar (60 psi)
  • 4.1 bar (60 psi)
  • Aplicación:
  • Gas line filter.
    Particulate removal from gases.
    Prefilters for suction.
  • Gas line filter.
    Particulate removal from gases.
    Prefilters for suction.
  • Gas line filter.
    Particulate removal from gases.
    Prefilters for suction.
  • Gas line filter.
    Particulate removal from gases.
    Prefilters for suction.
  • Material filtrante:
  • Laminated hydrophobically treated glass microfiber
  • Laminated hydrophobically treated glass microfiber
  • Laminated hydrophobically treated glass microfiber
  • Laminated hydrophobically treated glass microfiber