ContactUsLinkComponent
Whatman™ Lens Cleaning Tissue, Whatman products (Cytiva)
Whatman™ Lens Cleaning Tissue, Whatman products (Cytiva)
Catalog # 52846-108
Supplier:  Cytiva
Product available on GSA Advantage®
undefined
Whatman™ Lens Cleaning Tissue, Whatman products (Cytiva)
Catalog # 52846-108
Supplier:  Cytiva
Supplier Number:  2105-862
Product available on GSA Advantage®
Specifications
  • Descripción:
    Grade 105 lens cleaning tissue, sheet, 200×300 mm
  • Referencia:
    52846-108
  • Nº de proveedor:
    2105-862
Specifications
About this item
These cleaning tissues are suitable for use with lenses and any other optical surfaces made from glass, quartz or plastic.
  • Soft surface
  • Strong and highly absorbent
  • Chemically pure tissue free from silicones and other additives