Orders LinkContactUsLinkComponent
Heavy-Wall Flat-Bottom Boiling Flask, Kimble Chase
Heavy-Wall Flat-Bottom Boiling Flask, Kimble Chase
Heavy-Wall Flat-Bottom Boiling Flask, Kimble Chase
Certificates
Heavy-Wall Flat-Bottom Boiling Flask, Kimble Chase
Heavy-Wall Flat-Bottom Boiling Flask, Kimble Chase
2 Options Available Below
Product Details & Documents
Single neck flat bottom flask with a Standard Taper outer joint.
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Product Family Options
Product Information
  • Capacity
    Ext.Ø
    ST Joint
    Availability
    Price
  • 3 L
    185 mm
    29/42
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $5,552.06
    $5,552.06
    Each (1UN)
       $5,552.06
    Supplier Number: 601500-0929
  • 6 L
    234 mm
    24/40
    Log in for availability
    Log in for availability
    Case (1UN)
    Case (1UN) - $7,824.94
    $7,824.94
    Case (1UN)
       $7,824.94
    Supplier Number: 601500-1024