Orders LinkContactUsLinkComponent
VWR® Volumetric Pipettes, Class A, Serialized, Color-Coded
VWR® Volumetric Pipettes, Class A, Serialized, Color-Coded
VWR® Volumetric Pipettes, Class A, Serialized, Color-Coded
Supplier:  VWR International
Certificates
VWR® Volumetric Pipettes, Class A, Serialized, Color-Coded
VWR® Volumetric Pipettes, Class A, Serialized, Color-Coded
4 Options Available Below
Product Details & Documents
Made from Borosilicate 33 expansion glass, these pipettes are calibrated to deliver (TD) at 20 °C as per ASTM E969 standard.
  • Borosilicate glass fabrication
  • Color-coded for easy sorting
  • Individually serialized
Color-coded according to ASTM E1273 for easy sorting and selection. All pipettes come with an individual certificate of compliance. A unique three-digit number, along with the batch number, is printed on the pipette for Quality Assurance purposes.
Documents
Product Family Options
Información del producto
  • Capacity
    Band Color
    Tolerance
    Availability
    Price
  • 10124-360
    With individual certificate
    With individual certificate
    0.5 ml
    Black
    0.006 ml
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 3
    Paquete de 3 - $774.72
    $774.72
    Log in Para disponibilidad
    Case de 4 (3UN)
    Case de 4 (3UN) - $844.45
    $844.45
    Paquete de 3
       $774.72
    Case de 4 (3UN)
       $844.45
  • 10124-392
    With individual certificate
    With individual certificate
    2.5 ml
    0.01 ml
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 3
    Paquete de 3 - $706.47
    $706.47
    Log in Para disponibilidad
    Case de 4 (3UN)
    Case de 4 (3UN) - $770.04
    $770.04
    Paquete de 3
       $706.47
    Case de 4 (3UN)
       $770.04
  • 10124-004
    With individual certificate
    With individual certificate
    11 ml
    0.02 ml
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 3
    Paquete de 3 - $782.19
    $782.19
    Log in Para disponibilidad
    Case de 4 (3UN)
    Case de 4 (3UN) - $852.59
    $852.59
    Paquete de 3
       $782.19
    Case de 4 (3UN)
       $852.59
  • 10124-002
    With individual certificate
    With individual certificate
    12.5 ml
    Orange
    0.03 ml
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 3
    Paquete de 3 - $782.19
    $782.19
    Log in Para disponibilidad
    Case de 4 (3UN)
    Case de 4 (3UN) - $852.59
    $852.59
    Paquete de 3
       $782.19
    Case de 4 (3UN)
       $852.59